5,6-Dichloro-3-iodopyrazin-2-amine 95%

Supplier: Ambeed
Warning

A290604-100MG A290604-250MG A290604-1G
76648-016EA 62.6 USD
76648-016 76648-018 77748-563
5,6-Dichloro-3-iodopyrazin-2-amine 95%
5,6-Dichloro-3-iodopyrazin-2-amine

Precaución: for Research Use Only

Formula: C₄H₂Cl₂IN₃
MW: 289.8893 g/mol
MDL Number: MFCD29761095
CAS Number: 2089311-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR