Methyl 2-hydroxy-3-isopropylbenzoate 95%

Supplier: Ambeed
Warning

A290157-100MG A290157-1G A290157-250MG A290157-5G
76648-004EA 27 USD
76648-004 76648-006 76648-008 76648-010
Methyl 2-hydroxy-3-isopropylbenzoate 95%
Methyl 2-hydroxy-3-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MDL Number: MFCD24714020
CAS Number: 52159-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR