AG-881 99%

Supplier: Ambeed
Warning

A289383-10MG A289383-2MG A289383-50MG A289383-5MG A289383-1MG
76647-996EA 45 USD
76647-996 76647-998 76648-000 76648-002 77598-400
AG-881 99%
AG-881

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClF₆N₆
MDL Number: MFCD31630835
CAS Number: 1644545-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR