trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II) 98+%

Supplier: Ambeed
Warning

Synonyms: trans-Di-μ-acetatobis[2-[bis(2-methylphenyl)phosphino]benzyl]dipalladium, cataCXium® C

A288976-100MG A288976-1G A288976-250MG
76647-990EA 20 USD
76647-990 76647-992 76647-994
trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II) 98+%
trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II)

Precaución: for Research Use Only

Formula: C₄₆H₄₆O₄P₂Pd₂
MW: 937.6400 g/mol
MDL Number: MFCD01075746
CAS Number: 172418-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR