Belizatinib (TSR-011) 97%

Supplier: Ambeed
Warning

Synonyms: 4-fluoro-N-(6-((4-(2-hydroxypropan-2-yl)piperidin-1-yl)methyl)-1-((1s,4s)-4-(isopropylcarbamoyl)cyclohexyl)-1H-benzo[d]imidazol-2-yl)benzamide, 4-Fluoro-N-(6-((4-(2-hydroxypropan-2-yl)piperidin-1-yl)methyl)-1-(cis-4-(isopropylcarbamoyl)cyclohexyl)-1H-benzo[d]imidazol-2-yl)benzamide, Belizatinib, TSR011

A288219-100MG A288219-10MG A288219-1MG A288219-2MG A288219-50MG A288219-5MG
76647-972EA 350.1 USD
76647-972 76647-974 76647-976 76647-978 76647-980 76647-982
Belizatinib (TSR-011) 97%
Belizatinib (TSR-011)

Precaución: for Research Use Only

Formula: C₃₃H₄₄FN₅O₃
MW: 577.75 g/mol
MDL Number: MFCD31382200
CAS Number: 1357920-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR