LOXO-195 98%

Supplier: Ambeed
Warning

Synonyms: (3aR,10R)-5-Fluoro-1,2,3,3a,8,9,10,11-octahydro-10-methyl-12H-15,17-ethenopyrazolo[3,4-d]pyrido[2,3-k]pyrrolo[2,1-m][1,3,7]triazacyclotridecin-12-one, (13E,14E,22R,6R)-35-fluoro-6-methyl-7-aza-1(5,3)-pyrazolo[1,5-a]pyrimidina-3(3,2)-pyridina-2(1,2)-pyrrolidinacyclooctaphan-8-one, Selitrectinib

A285925-10MG A285925-1MG A285925-25MG A285925-5MG A285925-100MG A285925-50MG
76647-956EA 65.78 USD
76647-956 76647-958 76647-960 76647-962 77616-222 77616-224
LOXO-195 98%
LOXO-195

Precaución: for Research Use Only

Formula: C₂₀H₂₁FN₆O
MW: 380.43 g/mol
MDL Number: MFCD31620755
CAS Number: 2097002-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR