Ethyl 5-methyl-1H-pyrrole-3-carboxylate 97%

Supplier: Ambeed
Warning

A283978-100MG A283978-250MG
76647-950EA 79.57 USD
76647-950 76647-952
Ethyl 5-methyl-1H-pyrrole-3-carboxylate 97%
Ethyl 5-methyl-1H-pyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.1784 g/mol
MDL Number: MFCD01006756
CAS Number: 2199-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR