ω-Bromo-2,5-dimethoxyacetophenone 99%

Supplier: Ambeed
Danger

A279529-100G A279529-5G A279529-25G
76647-936EA 108 USD
76647-936 76647-938 77616-184
ω-Bromo-2,5-dimethoxyacetophenone 99%
ω-Bromo-2,5-dimethoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MDL Number: MFCD00000198
CAS Number: 1204-21-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR