6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydro-6,7-dimethoxyquinazoline-2,4-dione, 6,7-Dimethoxyquinazoline-2,4-dione

A279329-100G A279329-25G A279329-5G
76647-930EA 118.83 USD
76647-930 76647-932 76647-934
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione 98%
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₄
MW: 222.2 g/mol
MDL Number: MFCD00023889
CAS Number: 28888-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR