7-Fluoro-1H-indazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A276773-100MG A276773-250MG
76647-920EA 152.77 USD
76647-920 76647-922
7-Fluoro-1H-indazole-4-carboxylic acid 97%
7-Fluoro-1H-indazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MW: 180.1359 g/mol
MDL Number: MFCD31728867
CAS Number: 1783619-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR