5-Chloro-6-methoxynicotinaldehyde ≥99%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-5-formyl-2-methoxypyridine
A275596-250MG
A275596-100MG
76647-918EA
47
USD
76647-918
77616-154
5-Chloro-6-methoxynicotinaldehyde ≥99%
5-Chloro-6-methoxynicotinaldehyde
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₂ MW: 171.5810 g/mol |
MDL Number:
MFCD04972383 CAS Number: 132865-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...