(2-Bromo-4,5-dimethoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4,5-dimethoxyphenylacetic acid

A275063-1G A275063-5G
76647-914EA 56.23 USD
76647-914 76647-916
(2-Bromo-4,5-dimethoxyphenyl)acetic acid 97%
(2-Bromo-4,5-dimethoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₄
MW: 275.1 g/mol
MDL Number: MFCD00016819
CAS Number: 4697-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR