5-Methyl-1H-1,2,4-triazol-3-amine 95%

Supplier: Ambeed
Danger

A275060-250MG A275060-1G
76647-912EA 39 USD
76647-912 77598-392
5-Methyl-1H-1,2,4-triazol-3-amine 95%
5-Methyl-1H-1,2,4-triazol-3-amine

Precaución: for Research Use Only

Formula: C₃H₆N₄
MDL Number: MFCD00205267
CAS Number: 53741-81-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR