4-Chloro-4'-fluorobutyrophenone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-1-(4-fluorophenyl)butan-1-one, ω-Chloro-4-fluorobutyrophenone, γ-Chloro-4-fluorobutyrophenone

A268570-100G A268570-10G A268570-25G A268570-500G A268570-5G
76647-866EA 88.06 USD
76647-866 76647-868 76647-870 76647-872 76647-874
4-Chloro-4'-fluorobutyrophenone 97%
4-Chloro-4'-fluorobutyrophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO
MW: 200.64 g/mol
MDL Number: MFCD00001007
CAS Number: 3874-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR