3-Bromopropyl Acetate 95%

Supplier: Ambeed
Warning

A267787-1G A267787-250MG A267787-25G A267787-5G A267787-100G A267787-500G
76647-858EA 12 USD
76647-858 76647-860 76647-862 76647-864 77593-654 77748-510
3-Bromopropyl Acetate 95%
3-Bromopropyl Acetate

Precaución: for Research Use Only

Formula: C₅H₉BrO₂
MDL Number: MFCD02258489
CAS Number: 592-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR