(R)-1-(2,3-Difluorophenyl)ethanamine 97%

Supplier: Ambeed
Warning

A264942-100MG A264942-1G
76647-850EA 125.19 USD
76647-850 76647-852
(R)-1-(2,3-Difluorophenyl)ethanamine 97%
(R)-1-(2,3-Difluorophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₉F₂N
MW: 157.1606 g/mol
MDL Number: MFCD06761982
CAS Number: 1339886-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR