GSK-2018682 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4-(5-(5-chloro-6-isopropoxypyridin-3-yl)-1,2,4-oxadiazol-3-yl)-1H-indol-1-yl)butanoic acid, GSK2018682
A260586-10MG
A260586-5MG
76647-842EA
160.2
USD
76647-842
76647-844
GSK-2018682 97%
GSK-2018682
Precaución: for Research Use Only
Formula:
C₂₂H₂₁ClN₄O₄ MW: 440.88 g/mol |
MDL Number:
MFCD18633078 CAS Number: 1034688-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...