5-tert-Butyl 1-ethyl 5-azaspiro[2.4]heptane-1,5-dicarboxylate 98%

Supplier: Ambeed
Warning

A258642-100MG A258642-250MG
76647-834EA 219.61 USD
76647-834 77748-486
5-tert-Butyl 1-ethyl 5-azaspiro[2.4]heptane-1,5-dicarboxylate 98%
5-tert-Butyl 1-ethyl 5-azaspiro[2.4]heptane-1,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₄
MW: 269.3367 g/mol
MDL Number: MFCD28015471
CAS Number: 150543-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR