(4'-(tert-Butyl)-[1,1'-biphenyl]-2-yl)boronic acid 97%

Supplier: Ambeed
Warning

A257514-1G A257514-25G A257514-5G
76647-824EA 102.91 USD
76647-824 76647-826 76647-828
(4'-(tert-Butyl)-[1,1'-biphenyl]-2-yl)boronic acid 97%
(4'-(tert-Butyl)-[1,1'-biphenyl]-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₉BO₂
MW: 254.1300 g/mol
MDL Number: MFCD18383634
CAS Number: 1228458-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR