(6-Bromo-2-fluoropyridin-3-yl)methanol 95%

Supplier: Ambeed
Warning

A257504-100MG A257504-250MG A257504-5G A257504-1G
76647-818EA 44 USD
76647-818 76647-820 76647-822 77593-610
(6-Bromo-2-fluoropyridin-3-yl)methanol 95%
(6-Bromo-2-fluoropyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MDL Number: MFCD16607005
CAS Number: 1227563-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR