tert-butyl N-[trans-4-[(methanesulfonyloxy)methyl]cyclohexyl]carbamate 98%

Supplier: Ambeed
Warning

A256652-100MG A256652-1G A256652-250MG A256652-5G
76647-806EA 7 USD
76647-806 76647-808 76647-810 76647-812
tert-butyl N-[trans-4-[(methanesulfonyloxy)methyl]cyclohexyl]carbamate 98%
tert-butyl N-[trans-4-[(methanesulfonyloxy)methyl]cyclohexyl]carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO₅S
MDL Number: MFCD28383641
CAS Number: 683269-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR