Boc-L-octahydroindole-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Oic-OH, (2S,3aS,7aS)-1-(tert-Butoxycarbonyl)octahydro-1H-indole-2-carboxylic acid, Boc-(2S,3aS,7aS)-octahydro-1H-indole-2-carboxylic acid
A256462-10G
A256462-1G
A256462-5G
A256462-25G
A256462-100G
76647-800EA
41
USD
76647-800
76647-802
76647-804
77616-072
77748-478
Boc-L-octahydroindole-2-carboxylic acid 97%
Boc-L-octahydroindole-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₂₃NO₄ |
MDL Number:
MFCD09750486 CAS Number: 109523-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...