Pyrrolo[1,2-b]pyridazin-6-ylmethanol 97%

Supplier: Ambeed
Warning

A256329-100MG A256329-1G A256329-250MG A256329-50MG
76647-792EA 196 USD
76647-792 76647-794 76647-796 76647-798
Pyrrolo[1,2-b]pyridazin-6-ylmethanol 97%
Pyrrolo[1,2-b]pyridazin-6-ylmethanol

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD22396130
CAS Number: 1709825-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR