2-Dicyclohexylphosphino-2-(N,N-dimethylamino)biphenyl(2′-amino-1,1′-biphenyl-2-yl) palladium(II) 98%
Supplier: Ambeed
|
Warning
|
A255769-100MG
A255769-250MG
76647-780EA
68.96
USD
76647-780
76647-782
2-Dicyclohexylphosphino-2-(N,N-dimethylamino)biphenyl(2′-amino-1,1′-biphenyl-2-yl) palladium(II) 98%
2-Dicyclohexylphosphino-2-(N,N-dimethylamino)biphenyl(2′-amino-1,1′-biphenyl-2-yl) palladium(II)
Precaución: for Research Use Only
Formula:
C₃₈H₄₆ClN₂PPd MW: 703.63 g/mol |
MDL Number:
MFCD27978421 CAS Number: 2195390-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...