3-Fluoro-5-nitro-1-trifluoromethylbenzene 95%

Supplier: Ambeed
Warning

A1339484-100G A1339484-10G A1339484-1G A1339484-250MG A1339484-25G A1339484-5G
76657-998EA 892.22 USD
76657-998 76658-000 76658-002 76658-004 76658-006 76658-008
3-Fluoro-5-nitro-1-trifluoromethylbenzene 95%
3-Fluoro-5-nitro-1-trifluoromethylbenzene

Precaución: for Research Use Only

Formula: C7H3F4NO2
MW: 209.0978 g/mol
MDL Number: MFCD07779168
CAS Number: 454-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR