1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid xhydrochloride 98%

Supplier: Ambeed
Warning

A250946-100G A250946-25G
76647-760EA 16.98 USD
76647-760 76647-762
1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid xhydrochloride 98%
1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid xhydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₉ClFN₃O₃
MW: 367.802 g/mol
MDL Number: MFCD00079044
CAS Number: 86483-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR