5-(4-Isopropoxy-3-methylphenyl)-1H-pyrrole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A250493-100MG A250493-250MG
76647-756EA 60 USD
76647-756 76647-758
5-(4-Isopropoxy-3-methylphenyl)-1H-pyrrole-2-carboxylic acid 97%
5-(4-Isopropoxy-3-methylphenyl)-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₃
CAS Number: 1517011-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR