2-Amino-6-(trifluoromethyl)benzamide 95%

Supplier: Ambeed
Warning

A250187-100MG A250187-250MG
76647-752EA 75 USD
76647-752 76647-754
2-Amino-6-(trifluoromethyl)benzamide 95%
2-Amino-6-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₇F₃N₂O
CAS Number: 869936-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR