3-(1-Methyl-1H-pyrazol-3-yl)pyridin-2-amine 97%

Supplier: Ambeed
Warning

A1339464-100MG A1339464-250MG
76657-994EA 135.8 USD
76657-994 76657-996
3-(1-Methyl-1H-pyrazol-3-yl)pyridin-2-amine 97%
3-(1-Methyl-1H-pyrazol-3-yl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₉H₁₀N₄
MW: 174.2025 g/mol
MDL Number: MFCD22066611
CAS Number: 1368171-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR