2-Amino-3-(2-chloro-4-methylphenyl)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-Phe(2-Cl-4-Me)-OH

A1339350-1G A1339350-5G
76657-990EA 36.08 USD
76657-990 76657-992
2-Amino-3-(2-chloro-4-methylphenyl)propanoic acid 97%
2-Amino-3-(2-chloro-4-methylphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂
MW: 213.6608 g/mol
MDL Number: MFCD18697988
CAS Number: 1270310-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR