Pyrazolo[1,5-a]pyrimidin-5-amine 98%

Supplier: Ambeed
Warning

A249441-250MG A249441-50MG
76647-740EA 130 USD
76647-740 77748-464
Pyrazolo[1,5-a]pyrimidin-5-amine 98%
Pyrazolo[1,5-a]pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₆H₆N₄
MDL Number: MFCD19216038
CAS Number: 216066-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR