5,6,7,8-Tetrahydro-4H-pyrazolo[1,5-a][1,4]diazepine dihydrochloride 97%

Supplier: Ambeed
Warning

A1339306-100MG A1339306-1G A1339306-250MG
76657-984EA 102.91 USD
76657-984 76657-986 76657-988
5,6,7,8-Tetrahydro-4H-pyrazolo[1,5-a][1,4]diazepine dihydrochloride 97%
5,6,7,8-Tetrahydro-4H-pyrazolo[1,5-a][1,4]diazepine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃Cl₂N₃
MW: 210.1042 g/mol
MDL Number: MFCD34165673
CAS Number: 2682114-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR