(R)-5-Fluoro-N-(4-fluoro-3-(3-imino-2,5-dimethyl-1,1-dioxido-1,2,4-thiadiazinan-5-yl)phenyl)picolinamide 2,2,2-trifluoroacetate 95%

Supplier: Ambeed
Warning

A246701-100MG A246701-1G A246701-250MG
76647-728EA 128.37 USD
76647-728 76647-730 76647-732
(R)-5-Fluoro-N-(4-fluoro-3-(3-imino-2,5-dimethyl-1,1-dioxido-1,2,4-thiadiazinan-5-yl)phenyl)picolinamide 2,2,2-trifluoroacetate 95%
(R)-5-Fluoro-N-(4-fluoro-3-(3-imino-2,5-dimethyl-1,1-dioxido-1,2,4-thiadiazinan-5-yl)phenyl)picolinamide 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₁₉H₁₈F₅N₅O₅S
MW: 523.4337 g/mol
MDL Number: MFCD28963974
CAS Number: 2095432-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR