1-Ethyl-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A246698-10G A246698-1G A246698-250MG A246698-25G A246698-5G
76647-718EA 437.1 USD
76647-718 76647-720 76647-722 76647-724 76647-726
1-Ethyl-1H-pyrazole-5-carboxylic acid 95%
1-Ethyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD02090833
CAS Number: 400755-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR