3-Bromo-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A246145-100MG A246145-1G A246145-250MG A246145-5G
76647-710EA 52 USD
76647-710 76647-712 76647-714 76647-716
3-Bromo-1H-pyrazole-5-carboxylic acid 95%
3-Bromo-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O₂
MDL Number: MFCD22395739
CAS Number: 1905484-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR