Dabrafenib methanesulfonate salt ≥99%

Supplier: Ambeed
Warning

Synonyms: Dabrafenib Mesylate, N-(3-(5-(2-Aminopyrimidin-4-yl)-2-(tert-butyl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulphonamide methanesulphonate, GSK-2118436 mesylate

A244638-10MG A244638-1MG A244638-5MG A244638-25MG A244638-50MG A244638-250MG A244638-100MG A244638-1G
76647-704EA 20.16 USD
76647-704 76647-706 76647-708 77593-554 77593-556 77616-010 77748-431 77748-432
Dabrafenib methanesulfonate salt ≥99%
Dabrafenib methanesulfonate salt

Precaución: for Research Use Only

Formula: C₂₄H₂₄F₃N₅O₅S₃
MW: 615.67 g/mol
MDL Number: MFCD20922872
CAS Number: 1195768-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR