Methyl 2-amino-4-bromothiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

A243876-100MG A243876-250MG
76647-700EA 124.13 USD
76647-700 76647-702
Methyl 2-amino-4-bromothiophene-3-carboxylate 98%
Methyl 2-amino-4-bromothiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.0863 g/mol
MDL Number: MFCD25956968
CAS Number: 1239461-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR