3-(2-((tert-Butoxycarbonyl)amino)ethoxy)propanoic acid 95%
Supplier: Ambeed
|
Warning
|
A242736-100MG
A242736-1G
A242736-250MG
A242736-25G
A242736-5G
A242736-100G
76647-690EA
6.37
USD
76647-690
76647-692
76647-694
76647-696
76647-698
77616-002
3-(2-((tert-Butoxycarbonyl)amino)ethoxy)propanoic acid 95%
3-(2-((tert-Butoxycarbonyl)amino)ethoxy)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₅ MW: 233.2616 g/mol |
MDL Number:
MFCD22574796 CAS Number: 1260092-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...