3-Bromo-4-chloro-2-fluoropyridine 95%

Supplier: Ambeed
Warning

A240871-100MG A240871-250MG A240871-1G A240871-5G
76647-682EA 45 USD
76647-682 76647-684 77748-423 77748-424
3-Bromo-4-chloro-2-fluoropyridine 95%
3-Bromo-4-chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MDL Number: MFCD18257573
CAS Number: 1211528-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR