6-Bromo-2-ethyl-N,8-dimethylimidazo[1,2-a]pyridin-3-amine 95%

Supplier: Ambeed
Warning

A240610-100MG A240610-1G A240610-250MG
76647-676EA 58.35 USD
76647-676 76647-678 76647-680
6-Bromo-2-ethyl-N,8-dimethylimidazo[1,2-a]pyridin-3-amine 95%
6-Bromo-2-ethyl-N,8-dimethylimidazo[1,2-a]pyridin-3-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN₃
MW: 268.1530 g/mol
MDL Number: MFCD30803417
CAS Number: 1628264-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR