2,6-Diamino-9,10-anthraquinone 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Diaminoanthracene-9,10-dione

A1339134-25G A1339134-500G
76657-980EA 15.92 USD
76657-980 76657-982
2,6-Diamino-9,10-anthraquinone 95%
2,6-Diamino-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O₂
MW: 238.2414 g/mol
MDL Number: MFCD00001234
CAS Number: 131-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR