5-Nitro-1H-1,2,3-triazole 98%

Supplier: Ambeed
Danger

A234930-250MG A234930-100MG
76647-664EA 17 USD
76647-664 77748-404
5-Nitro-1H-1,2,3-triazole 98%
5-Nitro-1H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₂H₂N₄O₂
MDL Number: MFCD11502061
CAS Number: 14544-45-7
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR