(2,2'-Bipyridine)nickel(II) dibromide 97%

Supplier: Ambeed
Warning

A234394-1G A234394-5G A234394-10G A234394-25G A234394-100G
76647-652EA 12 USD
76647-652 76647-654 77622-270 77622-272 77783-370
(2,2'-Bipyridine)nickel(II) dibromide 97%
(2,2'-Bipyridine)nickel(II) dibromide

Precaución: for Research Use Only

Formula: C₁₀H₈Br₂N₂Ni
MW: 374.68 g/mol
MDL Number: MFCD31728660
CAS Number: 46389-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR