1-Decyl-3-methylimidazoliumbis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Warning

Synonyms: 1-Decyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide

A233557-25G A233557-5G
76647-648EA 59 USD
76647-648 76647-650
1-Decyl-3-methylimidazoliumbis(trifluoromethanesulfonyl)imide 98%
1-Decyl-3-methylimidazoliumbis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₆H₂₇F₆N₃O₄S₂
MW: 503.52 g/mol
MDL Number: MFCD12022376
CAS Number: 433337-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR