1,1'-(9-(2-(Isopropylamino)ethyl)-9H-carbazole-3,6-diyl)diethanone hydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: CBL0137 HCl

A232907-1MG A232907-2MG A232907-50MG A232907-5MG A232907-100MG A232907-1G
76647-640EA 28.65 USD
76647-640 76647-642 76647-644 76647-646 77748-397 77748-398
1,1'-(9-(2-(Isopropylamino)ethyl)-9H-carbazole-3,6-diyl)diethanone hydrochloride 99%
1,1'-(9-(2-(Isopropylamino)ethyl)-9H-carbazole-3,6-diyl)diethanone hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClN₂O₂
MW: 372.89 g/mol
MDL Number: MFCD28160457
CAS Number: 1197397-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR