Methyl 3-bromo-1H-pyrazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A232498-100MG A232498-1G A232498-250MG A232498-5G
76647-632EA 26.53 USD
76647-632 76647-634 76647-636 76647-638
Methyl 3-bromo-1H-pyrazole-5-carboxylate 97%
Methyl 3-bromo-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MW: 205.0094 g/mol
MDL Number: MFCD27987968
CAS Number: 2246373-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR