2-Bromo-6-chloro-1,7-naphthyridine 95%

Supplier: Ambeed
Warning

A231856-100MG A231856-250MG A231856-1G
76647-614EA 122 USD
76647-614 76647-616 77593-504
2-Bromo-6-chloro-1,7-naphthyridine 95%
2-Bromo-6-chloro-1,7-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MDL Number: MFCD09261002
CAS Number: 1246549-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR