(S)-Oxetan-2-ylmethanamine ≥95%, ee 99%

Supplier: Ambeed
Warning

A228426-100MG A228426-1G A228426-250MG A228426-5G
76647-602EA 20 USD
76647-602 76647-604 76647-606 76647-608
(S)-Oxetan-2-ylmethanamine ≥95%, ee 99%
(S)-Oxetan-2-ylmethanamine

Precaución: for Research Use Only

Formula: C₄H₉NO
MDL Number: MFCD31727897
CAS Number: 2091328-57-1

Order Now

Specification Test Results

Loss on drying ≤ 2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR