(3-Bromo-5-(tert-butyl)phenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-5-tert-butylphenylboronic acid
A227918-250MG
A227918-5G
A227918-25G
A227918-1G
76647-598EA
14.86
USD
76647-598
76647-600
77615-906
77748-383
(3-Bromo-5-(tert-butyl)phenyl)boronic acid 98%
(3-Bromo-5-(tert-butyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₄BBrO₂ MW: 256.9320 g/mol |
MDL Number:
MFCD28167195 CAS Number: 1373881-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...