1-(2-Bromobenzyl)-1H-pyrrole-2,5-dione 95%

Supplier: Ambeed
Warning

A226293-100MG A226293-1G A226293-250MG
76647-588EA 10.61 USD
76647-588 76647-590 76647-592
1-(2-Bromobenzyl)-1H-pyrrole-2,5-dione 95%
1-(2-Bromobenzyl)-1H-pyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO₂
MW: 266.0907 g/mol
CAS Number: 140480-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR